missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentyl-L-alanine, 95%

CAS: 99295-82-6 | C8H15NO2 | 157.213 g/mol

Supplier:  Thermo Scientific Chemicals H6660003

Quantity 1 g

 View more versions of this product

Catalog No. AAH6660003


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

99295-82-6
157.213
KDYAKYRBGLKMAK-ZETCQYMHSA-N
7000137
C1CCC(C1)CC(C(=O)O)N
C8H15NO2
MFCD01632069
s-2-amino-3-cyclopentylpropanoic acid, h-cpa-oh, 3-cyclopentylalanine, l-3-cyclopentylalanine, 3-cyclopentane-l-alanine, 2s-2-amino-3-cyclopentylpropanoic acid, s-2-amino-3-cyclopentyl-propionic acid, s-alfa-amino-cyclopentanepropanoic acid ;3-cyclopentane-l-alanine, ambotzhaa1075, l-beta-cyclopentylalanine
(2S)-2-amino-3-cyclopentylpropanoic acid

Specifications

3-Cyclopentyl-L-alanine
White
MFCD01632069
s-2-amino-3-cyclopentylpropanoic acid, h-cpa-oh, 3-cyclopentylalanine, l-3-cyclopentylalanine, 3-cyclopentane-l-alanine, 2s-2-amino-3-cyclopentylpropanoic acid, s-2-amino-3-cyclopentyl-propionic acid, s-alfa-amino-cyclopentanepropanoic acid ;3-cyclopentane-l-alanine, ambotzhaa1075, l-beta-cyclopentylalanine
C1CCC(C1)CC(C(=O)O)N
1 g
7000137
95%
99295-82-6
C8H15NO2
10659781
KDYAKYRBGLKMAK-ZETCQYMHSA-N
(2S)-2-amino-3-cyclopentylpropanoic acid
157.213
157.21
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions