accessibility menu, dialog, popup

UserName

OOHH2N

CAS RN 99295-82-6

IUPAC Name: 2-amino-3-cyclopentylpropanoic acid
Synonyms: 2-amino-3-cyclopentylpropanoic acid
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: KDYAKYRBGLKMAK-UHFFFAOYNA-N
SMILES: NC(CC1CCCC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)