missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentene-1-carboxylic Acid 97.0+%, TCI America™

Catalog No. C19145G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19145G 5 g
C19141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19145G Supplier TCI America Supplier No. C19145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7686-77-3
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00101764
InChI Key XVSYDLITVYBCBD-UHFFFAOYSA-N
Synonym 3-cyclopentene-1-carboxylic acid, cyclopent-3-enecarboxylic acid, 3-cyclopentenecarboxylic acid, 3-cyclopentene-1-carhoxylic acid, cyclopent-3-ene carboxylic acid, cyclopentene-3-carboxylic acid, ghl.pd_mitscher_leg0.243, 3-cyclopentene-1-carboxylicacid, acmc-1bh22, ksc377a8b
PubChem CID 853756
IUPAC Name cyclopent-3-ene-1-carboxylic acid
SMILES C1C=CCC1C(=O)O

Specifications

Color Yellow
Boiling Point 215°C
Quantity 5 g
UN Number 3265
Formula Weight 112.13
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Cyclopentene-1-carboxylic Acid