accessibility menu, dialog, popup

UserName

CAS RN 7686-77-3

IUPAC Name: cyclopent-3-ene-1-carboxylate
Synonyms: cyclopent-3-ene-1-carboxylate
Molecular Weight (g/mol): 111.12
Molecular Formula: C6H7O2
InChi Key: XVSYDLITVYBCBD-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1CC=CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results