missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopenten-1-ol

Catalog No. AC467550010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467550010 1 g
AC467550050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467550010 Supplier Thermo Scientific Chemicals Supplier No. 467550010

CAS: 14320-38-8 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14320-38-8
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00798186
InChI Key WEIMJSIRDZDHAH-UHFFFAOYSA-N
IUPAC Name cyclopent-3-en-1-ol
SMILES OC1CC=CC1

Specifications

Boiling Point 136.0°C
Flash Point 42°C
Quantity 1 g
Formula Weight 84.13
Chemical Name or Material 3-Cyclopenten-1-ol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Flammable liquid and vapor.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only