accessibility menu, dialog, popup

UserName

OH

CAS RN 14320-38-8

IUPAC Name: cyclopent-3-en-1-ol
Synonyms: cyclopent-3-en-1-ol
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: WEIMJSIRDZDHAH-UHFFFAOYSA-N
SMILES: OC1CC=CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results