missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexylamino-2-hydroxypropanesulfonic Acid Sodium Salt 95.0+%, TCI America™

Catalog No. c156225g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C156225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C156225G Supplier TCI America Supplier No. C156225G
Only null left

Chemical Identifiers

CAS 102601-34-3
Molecular Formula C9H12NO4S
Molecular Weight (g/mol) 230.26
MDL Number MFCD00070063
InChI Key CLEVULOKVPBHCU-VIFPVBQESA-M
Synonym capso sodium salt, sodium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate, 3-cyclohexylamino-2-hydroxypropanesulfonic acid sodium salt, capso sodium, capso, 0.2m buffer solution, ph 8.5, capso, 0.2m buffer solution, ph 9.0, capso, 0.2m buffer solution, ph 9.5, sodium 3-cyclohexylamino-2-hydroxypropanesulfonate, sodium 3-cyclohexylamino-2-hydroxy-propane-1-sulfonate, potassium 3-cyclohexylamino-2-hydroxypropane-1-sulfonate
PubChem CID 25000353
IUPAC Name (2S)-2-hydroxy-3-(phenylamino)propane-1-sulfonate
SMILES O[C@@H](CNC1=CC=CC=C1)CS([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 259.30
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyclohexylamino-2-hydroxypropanesulfonic Acid Sodium Salt