missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexyl-1-propanol 99.0+%, TCI America™

Catalog No. p-7124831
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11665ML 5 mL
C116625ML 25 mL
C1166100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C11665ML Supplier TCI America Supplier No. C11665ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1124-63-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00001528
InChI Key CLYAQFSQLQTVNO-UHFFFAOYSA-N
Synonym cyclohexanepropanol, 3-cyclohexyl-1-propanol, 3-cyclohexylpropanol, unii-kjb3vo906f, kjb3vo906f, cyclohexylpropyl alcohol, 3-hydroxypropylcyclohexane, 3-cyclohexylpropyl alcohol, acmc-1c5ti
PubChem CID 70763
IUPAC Name 3-cyclohexylpropan-1-ol
SMILES C1CCC(CC1)CCCO

Specifications

Color Colorless
Boiling Point 223°C
Quantity 5 mL
Formula Weight 142.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Cyclohexyl-1-propanol