accessibility menu, dialog, popup

UserName

HO

CAS RN 1124-63-6

IUPAC Name: 3-cyclohexylpropan-1-ol
Synonyms: cyclohexylpropanol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: CLYAQFSQLQTVNO-UHFFFAOYSA-N
SMILES: OCCCC1CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)