missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexene-1-methanol 98.0+%, TCI America™

Catalog No. p-7125460
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C243525G 25 g
C2435250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C243525G Supplier TCI America Supplier No. C243525G
Only null left

Chemical Identifiers

CAS 1679-51-2
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001574
InChI Key VEIYJWQZNGASMA-UHFFFAOYNA-N
Synonym 1,2,3,6-Tetrahydrobenzyl Alcohol
PubChem CID 15512
IUPAC Name (cyclohex-3-en-1-yl)methanol
SMILES OCC1CCC=CC1

Specifications

Color Colorless
Boiling Point 100°C
Quantity 25 g
Formula Weight 112.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Cyclohexene-1-methanol