CAS RN 1679-51-2
CAS RN 1679-51-2
IUPAC Name:
(cyclohex-3-en-1-yl)methanol
Synonyms:
cyclohex-3-en-1-ylmethanol
Molecular Weight (g/mol):
112.17
Molecular Formula:
C7H12O
InChi Key:
VEIYJWQZNGASMA-UHFFFAOYNA-N
SMILES:
OCC1CCC=CC1
1
–
1
of
1
results
1
3-Cyclohexene-1-methanol 98.0+%, TCI America™
CAS: 1679-51-2 Molecular Formula: C7H12O Molecular Weight (g/mol): 112.17 MDL Number: MFCD00001574 InChI Key: VEIYJWQZNGASMA-UHFFFAOYNA-N Synonym: 1,2,3,6-Tetrahydrobenzyl Alcohol PubChem CID: 15512 IUPAC Name: (cyclohex-3-en-1-yl)methanol SMILES: OCC1CCC=CC1