missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine 98.0+%, TCI America™

Catalog No. p-7124433
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C045625G 25 g
C0456500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C045625G Supplier TCI America Supplier No. C045625G
Only null left

Chemical Identifiers

CAS 100-54-9
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00006372
InChI Key GZPHSAQLYPIAIN-UHFFFAOYSA-N
Synonym 3-cyanopyridine, nicotinonitrile, 3-pyridinecarbonitrile, nicotinic acid nitrile, 3-pyridinenitrile, 3-pyridylcarbonitrile, 3-azabenzonitrile, 3-cyjanopirydyna, nitryl kwasu nikotynowego, 3-cyjanopirydyna polish
PubChem CID 79
ChEBI CHEBI:86556
IUPAC Name pyridine-3-carbonitrile
SMILES N#CC1=CC=CN=C1

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 201°C
Quantity 25 g
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyanopyridine
EINECSNumber (5)-0742
RTECSNumber QT3030000
TSCA Yes