missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine, 98%

Catalog No. AC110860025
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110860025 2.5 kg Glass bottle
AC110860050 5 g Glass bottle
AC110861000 100 g Glass bottle
AC110865000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC110860025 Supplier Thermo Scientific Chemicals Supplier No. 110860025
Only null left

CAS: 100-54-9 | C6H4N2 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-54-9
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00006372
InChI Key GZPHSAQLYPIAIN-UHFFFAOYSA-N
Synonym 3-cyanopyridine, nicotinonitrile, 3-pyridinecarbonitrile, nicotinic acid nitrile, 3-pyridinenitrile, 3-pyridylcarbonitrile, 3-azabenzonitrile, 3-cyjanopirydyna, nitryl kwasu nikotynowego, 3-cyjanopirydyna polish
PubChem CID 79
ChEBI CHEBI:86556
IUPAC Name pyridine-3-carbonitrile
SMILES N#CC1=CC=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 47.0°C to 51.0°C
Color Colorless or White to Yellow
Density 1.1590g/mL
Boiling Point 201.0°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)CN
Quantity 2.5 kg
Beilstein 22, 41
Specific Gravity 1.159
Solubility Information Solubility in water: 140g/L (20°C)
Formula Weight 104.11
Percent Purity 98%
Physical Form Crystalline Mass, Crystals or Chunks
Chemical Name or Material 3-Cyanopyridine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-863-0
RTECSNumber QT3030000
TSCA TSCA

RUO – Research Use Only