missing translation for 'onlineSavingsMsg'
Learn More

(3-Cyanopropyl)triethoxysilane, 97%

Catalog No. AAL0536722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0536722 100 g
AAL0536714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0536722 Supplier Thermo Scientific Chemicals Supplier No. L0536722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1067-47-6 | C10H21NO3Si | 231.367 g/mol

(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Cyanopropyl)triethoxysilane is used as cyanoalkyl silane coupling agent.

Solubility
Not miscible in water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and water.

Chemical Identifiers

CAS 1067-47-6
Molecular Formula C10H21NO3Si
Molecular Weight (g/mol) 231.367
MDL Number MFCD00042636
InChI Key VGIURMCNTDVGJM-UHFFFAOYSA-N
Synonym 3-cyanopropyltriethoxysilane, butanenitrile, 4-triethoxysilyl, 4-triethoxysilyl butanenitrile, 4-triethoxysilyl butyronitrile, butyronitrile, 4-triethoxysilyl, silane, triethoxy 3-cyanopropyl, triethoxy-3-kyanpropylsilan, unii-n78w6a24dx, 3-cyanopropyl triethoxysilane, triethoxy-3-kyanpropylsilan czech
PubChem CID 14024
IUPAC Name 4-triethoxysilylbutanenitrile
SMILES CCO[Si](CCCC#N)(OCC)OCC

Specifications

Boiling Point 238°C to 241°C
Quantity 100 g
Flash Point 74°C (165°F)
Refractive Index 1.417
Sensitivity Moisture sensitive
Density 0.961
Linear Formula NCCH2CH2CH2Si(OCH2CH3)3
Beilstein 1942237
Solubility Information Not miscible in water.
Formula Weight 231.37
Percent Purity 97%
Chemical Name or Material (3-Cyanopropyl)triethoxysilane
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 213-931-4
RTECSNumber VV2785000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only