missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyanobenzenesulfonyl Chloride 98.0+%, TCI America™
$285.21 - $1137.08
Chemical Identifiers
| CAS | 56542-67-7 |
|---|---|
| Molecular Formula | C7H4ClNO2S |
| Molecular Weight (g/mol) | 201.624 |
| InChI Key | BHNRGBRMCNHNQD-UHFFFAOYSA-N |
| Synonym | 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463 |
| PubChem CID | 2801360 |
| IUPAC Name | 3-cyanobenzenesulfonyl chloride |
| SMILES | C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N |
Chemical Identifiers
| 56542-67-7 | |
| 201.624 | |
| 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463 | |
| 3-cyanobenzenesulfonyl chloride |
| C7H4ClNO2S | |
| BHNRGBRMCNHNQD-UHFFFAOYSA-N | |
| 2801360 | |
| C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N |
Specifications
| 56542-67-7 | |
| 111°C | |
| 1 g | |
| 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463 | |
| C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N | |
| 201.624 | |
| 201.62 | |
| Crystalline Powder |
| 51°C | |
| C7H4ClNO2S | |
| 2928 | |
| BHNRGBRMCNHNQD-UHFFFAOYSA-N | |
| 3-cyanobenzenesulfonyl chloride | |
| 2801360 | |
| ≥98.0% (GC,T) | |
| 3-Cyanobenzenesulfonyl Chloride |
Safety and Handling
TSCA : No