accessibility menu, dialog, popup

UserName

ClSOON

CAS RN 56542-67-7

IUPAC Name: 3-cyanobenzene-1-sulfonyl chloride
Synonyms: 3-cyanobenzenesulfonyl chloride
Molecular Weight (g/mol): 201.62
Molecular Formula: C7H4ClNO2S
InChi Key: BHNRGBRMCNHNQD-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CC(=C1)C#N
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)