missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. C18121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18121G 1 g
C18125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18121G Supplier TCI America Supplier No. C18121G

Chemical Identifiers

CAS 56542-67-7
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.624
InChI Key BHNRGBRMCNHNQD-UHFFFAOYSA-N
Synonym 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463
PubChem CID 2801360
IUPAC Name 3-cyanobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N

Specifications

Melting Point 51°C
Boiling Point 111°C
Quantity 1 g
UN Number 2928
Formula Weight 201.62
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyanobenzenesulfonyl Chloride