Learn More
3-Cyanobenzenesulfonyl chloride, 97%
CAS: 56542-67-7 | C7H4ClNO2S | 201.624 g/mol
$145.81 - $430.74
Chemical Identifiers
| CAS | 56542-67-7 |
|---|---|
| Molecular Formula | C7H4ClNO2S |
| Molecular Weight (g/mol) | 201.624 |
| MDL Number | MFCD00203480 |
| InChI Key | BHNRGBRMCNHNQD-UHFFFAOYSA-N |
| Synonym | 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463 |
| PubChem CID | 2801360 |
| IUPAC Name | 3-cyanobenzenesulfonyl chloride |
| SMILES | C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL2016903
|
Thermo Scientific Chemicals
L2016903 |
1 g |
Each for $145.81
|
|
|||||
|
AAL2016906
|
Thermo Scientific Chemicals
L2016906 |
5 g |
Each for $430.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56542-67-7 | |
| 201.624 | |
| BHNRGBRMCNHNQD-UHFFFAOYSA-N | |
| 2801360 | |
| C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N |
| C7H4ClNO2S | |
| MFCD00203480 | |
| 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463 | |
| 3-cyanobenzenesulfonyl chloride |
Specifications
| 56542-67-7 | |
| 114°C to 116°C (0.2 mmHg) | |
| C7H4ClNO2S | |
| 1 g | |
| 2832925 | |
| 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463 | |
| C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N | |
| 201.624 | |
| 201.63 | |
| 3-Cyanobenzenesulfonyl chloride |
| 47°C to 52°C | |
| >110°C (230°F) | |
| MFCD00203480 | |
| UN3261 | |
| Moisture sensitive | |
| BHNRGBRMCNHNQD-UHFFFAOYSA-N | |
| 3-cyanobenzenesulfonyl chloride | |
| 2801360 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only