missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzenesulfonyl chloride, 97%

Catalog No. AAL2016906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2016906 5 g
AAL2016903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2016906 Supplier Thermo Scientific Chemicals Supplier No. L2016906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56542-67-7 | C7H4ClNO2S | 201.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56542-67-7
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.624
MDL Number MFCD00203480
InChI Key BHNRGBRMCNHNQD-UHFFFAOYSA-N
Synonym 3-cyanobenzene-1-sulfonyl chloride, 3-cyanobenzene-1-sulfonylchloride, 3-cyano-benzenesulfonyl chloride, 3-cyanobenzenesulphonyl chloride, benzenesulfonyl chloride, 3-cyano, pubchem5108, acmc-1au46, 3-cyanobenzenesulfonylchloride, ksc269e7j, akos bb-9463
PubChem CID 2801360
IUPAC Name 3-cyanobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)C#N

Specifications

Melting Point 47°C to 52°C
Boiling Point 114°C to 116°C (0.2 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 2832925
Sensitivity Moisture sensitive
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 3-Cyanobenzenesulfonyl chloride
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only