missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Cyanoacetophenone 98.0+%, TCI America™
$59.69 - $200.21
Chemical Identifiers
| CAS | 6136-68-1 |
|---|---|
| Molecular Formula | C9H7NO |
| Molecular Weight (g/mol) | 145.16 |
| MDL Number | MFCD00001806 |
| InChI Key | SBCFGFDAZCTSRH-UHFFFAOYSA-N |
| Synonym | m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649 |
| PubChem CID | 80222 |
| IUPAC Name | 3-acetylbenzonitrile |
| SMILES | CC(=O)C1=CC=CC(=C1)C#N |
Chemical Identifiers
| 6136-68-1 | |
| 145.16 | |
| SBCFGFDAZCTSRH-UHFFFAOYSA-N | |
| 80222 | |
| CC(=O)C1=CC=CC(=C1)C#N |
| C9H7NO | |
| MFCD00001806 | |
| m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649 | |
| 3-acetylbenzonitrile |
Specifications
| 6136-68-1 | |
| 120°C | |
| MFCD00001806 | |
| 3439 | |
| SBCFGFDAZCTSRH-UHFFFAOYSA-N | |
| 3-acetylbenzonitrile | |
| 80222 | |
| ≥98.0% (GC) | |
| 3′-Cyanoacetophenone |
| 99°C | |
| C9H7NO | |
| 5 g | |
| m-cyanoacetophenone, 3'-cyanoacetophenone, benzonitrile, 3-acetyl, 3-cyanoacetophenone, 3-acetyl-benzonitrile, unii-tsj72034p9, 3-acetylbenzenecarbonitrile, m-acetylbenzonitrile, 3-acetyl benzonitrile, pubchem12649 | |
| CC(=O)C1=CC=CC(=C1)C#N | |
| 145.16 | |
| 145.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No