accessibility menu, dialog, popup

UserName

CH3ON

CAS RN 6136-68-1

IUPAC Name: 3-acetylbenzonitrile
Synonyms: M-cyanoacetophenone
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: SBCFGFDAZCTSRH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(=C1)C#N
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)