missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyano-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6206603
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 3-Cyano-5-fluorobenzeneboronic acid pinacol ester | |
| C13H15BFNO2 | |
| 1 g | |
| WWDTYZLFIFQZDE-UHFFFAOYSA-N | |
| 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile | |
| 46738194 | |
| 96% |
| 935685-88-4 | |
| MFCD11521351 | |
| 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 3-cyano-5-fluorobenzeneboronic acid pinacol ester, 3-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-5-fluorophenylboronic acid pinacol ester, 3-fluoro-5-cyanophenylboronic acid pinacol ester, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3.2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenecarbonitrile, benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
| CC1(C)OB(OC1(C)C)C1=CC(=CC(F)=C1)C#N | |
| 247.08 | |
| 247.07 | |
| Solid |
Chemical Identifiers
| 935685-88-4 | |
| 247.08 | |
| WWDTYZLFIFQZDE-UHFFFAOYSA-N | |
| 46738194 | |
| CC1(C)OB(OC1(C)C)C1=CC(=CC(F)=C1)C#N |
| C13H15BFNO2 | |
| MFCD11521351 | |
| 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 3-cyano-5-fluorobenzeneboronic acid pinacol ester, 3-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-5-fluorophenylboronic acid pinacol ester, 3-fluoro-5-cyanophenylboronic acid pinacol ester, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3.2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenecarbonitrile, benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
| 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures