missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. p-7054129
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62066MD 250 mg
AAH6206603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62066MD Supplier Thermo Scientific Chemicals Supplier No. H62066MD
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 935685-88-4
Molecular Formula C13H15BFNO2
Molecular Weight (g/mol) 247.08
MDL Number MFCD11521351
InChI Key WWDTYZLFIFQZDE-UHFFFAOYSA-N
Synonym 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 3-cyano-5-fluorobenzeneboronic acid pinacol ester, 3-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-5-fluorophenylboronic acid pinacol ester, 3-fluoro-5-cyanophenylboronic acid pinacol ester, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3.2-dioxaborolan-2-yl benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenecarbonitrile, benzonitrile, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 46738194
IUPAC Name 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC(=CC(F)=C1)C#N

Specifications

Quantity 250 mg
Formula Weight 247.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Cyano-5-fluorobenzeneboronic acid pinacol ester
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures