Learn More
3-Chromanecarboxylic acid, 97%, Thermo Scientific™
Identifiants chimiques
| CAS | 115822-57-6 |
|---|---|
| Molecular Formula | C10H10O3 |
| Molecular Weight (g/mol) | 178.187 |
| MDL Number | MFCD03783554 |
| InChI Key | UGAGZMGJJFSKQM-UHFFFAOYSA-N |
| Synonym | chroman-3-carboxylic acid, 3-chromanecarboxylic acid, chromane-3-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylic acid, 3,4-dihydro, 2h-1-benzopyran-3-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-benzopyran-3-carboxylic acid |
| PubChem CID | 2795458 |
| IUPAC Name | 3,4-dihydro-2H-chromene-3-carboxylic acid |
| SMILES | C1C(COC2=CC=CC=C21)C(=O)O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|
CC25801CB
|
N/A |
N/A
|
N/A | ||||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
CC25801DA
|
N/A |
N/A
|
N/A | ||||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
CC25801EA
|
N/A |
N/A
|
N/A | ||||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Identifiants chimiques
| 115822-57-6 | |
| 178.187 | |
| UGAGZMGJJFSKQM-UHFFFAOYSA-N | |
| 2795458 | |
| C1C(COC2=CC=CC=C21)C(=O)O |
| C10H10O3 | |
| MFCD03783554 | |
| chroman-3-carboxylic acid, 3-chromanecarboxylic acid, chromane-3-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylic acid, 3,4-dihydro, 2h-1-benzopyran-3-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-benzopyran-3-carboxylic acid | |
| 3,4-dihydro-2H-chromene-3-carboxylic acid |
Spécifications
| 115822-57-6 | |
| 100.0 | |
| C10H10O3 | |
| chroman-3-carboxylic acid, 3-chromanecarboxylic acid, chromane-3-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylic acid, 3,4-dihydro, 2h-1-benzopyran-3-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-benzopyran-3-carboxylic acid | |
| C1C(COC2=CC=CC=C21)C(=O)O | |
| 178.187 | |
| 178.19 | |
| 3-Chromanecarboxylic acid |
| 96.5 | |
| 97% | |
| MFCD03783554 | |
| UGAGZMGJJFSKQM-UHFFFAOYSA-N | |
| 3,4-dihydro-2H-chromene-3-carboxylic acid | |
| 2795458 | |
| 97% |
Sécurité et manipulation
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.