missing translation for 'onlineSavingsMsg'
Learn More

3-Chromanecarboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC25801EA

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC25801EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

115822-57-6
178.187
UGAGZMGJJFSKQM-UHFFFAOYSA-N
2795458
C1C(COC2=CC=CC=C21)C(=O)O
C10H10O3
MFCD03783554
chroman-3-carboxylic acid, 3-chromanecarboxylic acid, chromane-3-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylic acid, 3,4-dihydro, 2h-1-benzopyran-3-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-benzopyran-3-carboxylic acid
3,4-dihydro-2H-chromene-3-carboxylic acid

Specifications

3-Chromanecarboxylic acid
96.5
97%
C10H10O3
MFCD03783554
UGAGZMGJJFSKQM-UHFFFAOYSA-N
3,4-dihydro-2H-chromene-3-carboxylic acid
2795458
97%
115822-57-6
100.0
Amber Glass Bottle
10g
chroman-3-carboxylic acid, 3-chromanecarboxylic acid, chromane-3-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylic acid, 3,4-dihydro, 2h-1-benzopyran-3-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-benzopyran-3-carboxylic acid
C1C(COC2=CC=CC=C21)C(=O)O
178.187
178.19
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents