missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Catalog No. AAB2380606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2380606 5 g
AAB2380614 25 g
AAB2380622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2380606 Supplier Thermo Scientific Chemicals Supplier No. B2380606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2039-85-2 | C8H7Cl | 138.594 g/mol

It has been identified as a potential candidate for an electron resist to be used in direct device fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been identified as a potential candidate for an electron resist to be used in direct device fabrication.

Solubility
Not miscible or difficult to mix in water. Soluble in ether, and ethanol

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.

Chemical Identifiers

CAS 2039-85-2
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.594
MDL Number MFCD00000598
InChI Key BOVQCIDBZXNFEJ-UHFFFAOYSA-N
Synonym 3-chlorostyrene, 1-chloro-3-vinylbenzene, m-chlorostyrene, benzene, 1-chloro-3-ethenyl, styrene, m-chloro, unii-11k8g759hc, 1-chloro-3-vinyl-benzene, styrene, 3-chloro, meta-chlorostyrene, zlchem 318
PubChem CID 14905
IUPAC Name 1-chloro-3-ethenylbenzene
SMILES C=CC1=CC(=CC=C1)Cl

Specifications

Density 1.09
Boiling Point 188°C to 189°C
Flash Point 67°C (153°F)
Odor Characteristic
Refractive Index 1.562
Quantity 5 g
Beilstein 2038489
Solubility Information Not miscible or difficult to mix in water. Soluble in ether,and ethanol
Formula Weight 138.6
Percent Purity 98%
Chemical Name or Material 3-Chlorostyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 218-024-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only