missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyltriethoxysilane 97.0+%, TCI America™

Catalog No. p-7136683
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
T125325ML 25 mL
T1253100ML 100 mL
T1253500ML 500 mL
3 options

Catalog No. T125325ML

Supplier: TCI America T125325ML

Only null left

Chemical Identifiers

CAS 5089-70-3
Molecular Formula C9H21ClO3Si
Molecular Weight (g/mol) 240.799
MDL Number MFCD00018985
InChI Key KSCAZPYHLGGNPZ-UHFFFAOYSA-N
Synonym 3-chloropropyl triethoxysilane, 3-chloropropyl triethoxy silane, silane, 3-chloropropyl triethoxy, unii-x7rb20518m, triethoxy gamma-chloropropyl silane, gamma-chloropropyltriethoxysilane, triethoxy .gamma.-chloropropyl silane, chloropropyl triethoxysilane, dynasylan cpteo
PubChem CID 78771
IUPAC Name 3-chloropropyl(triethoxy)silane
SMILES CCO[Si](CCCCl)(OCC)OCC

Specifications

Color Yellow
Boiling Point 102°C
Quantity 25 mL
Formula Weight 240.80
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropropyltriethoxysilane
EINECSNumber (2)-2079
TSCA Yes