accessibility menu, dialog, popup

UserName

CAS RN 5089-70-3

IUPAC Name: (3-chloropropyl)triethoxysilane
Synonyms: (3-chloropropyl)triethoxysilane
Molecular Weight (g/mol): 240.8
Molecular Formula: C9H21ClO3Si
InChi Key: KSCAZPYHLGGNPZ-UHFFFAOYSA-N
SMILES: CCO[Si](CCCCl)(OCC)OCC
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)