missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyltriethoxysilane 97.0+%, TCI America™

Catalog No. T125325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T125325ML 25 mL
T1253100ML 100 mL
T1253500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T125325ML Supplier TCI America Supplier No. T125325ML

Chemical Identifiers

CAS 5089-70-3
Molecular Formula C9H21ClO3Si
Molecular Weight (g/mol) 240.799
MDL Number MFCD00018985
InChI Key KSCAZPYHLGGNPZ-UHFFFAOYSA-N
Synonym 3-chloropropyl triethoxysilane, 3-chloropropyl triethoxy silane, silane, 3-chloropropyl triethoxy, unii-x7rb20518m, triethoxy gamma-chloropropyl silane, gamma-chloropropyltriethoxysilane, triethoxy .gamma.-chloropropyl silane, chloropropyl triethoxysilane, dynasylan cpteo
PubChem CID 78771
IUPAC Name 3-chloropropyl(triethoxy)silane
SMILES CCO[Si](CCCCl)(OCC)OCC

Specifications

Color Yellow
Boiling Point 102°C
Quantity 25 mL
Formula Weight 240.80
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropropyltriethoxysilane
EINECSNumber (2)-2079
TSCA Yes