missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyldichloromethylsilane 97.0+%, TCI America™

$146.31 - $415.84

Chemical Identifiers

CAS 7787-93-1
Molecular Formula C4H9Cl3Si
Molecular Weight (g/mol) 191.551
MDL Number MFCD00000996
InChI Key UCJHMXXKIKBHQP-UHFFFAOYSA-N
Synonym 3-(Dichloromethylsilyl)propyl Chloride
PubChem CID 82235
IUPAC Name dichloro-(3-chloropropyl)-methylsilane
SMILES C[Si](CCCCl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D19955ML
View Documents
TCI America
D19955ML
5 mL
Each for $146.31
Only null left
 
D199525ML
View Documents
TCI America
D199525ML
25 mL
Each for $415.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7787-93-1
191.551
UCJHMXXKIKBHQP-UHFFFAOYSA-N
82235
C[Si](CCCCl)(Cl)Cl
C4H9Cl3Si
MFCD00000996
3-(Dichloromethylsilyl)propyl Chloride
dichloro-(3-chloropropyl)-methylsilane

Specifications

7787-93-1
C4H9Cl3Si
5 mL
3-(Dichloromethylsilyl)propyl Chloride
C[Si](CCCCl)(Cl)Cl
191.551
191.55
Liquid
185°C
MFCD00000996
2986
UCJHMXXKIKBHQP-UHFFFAOYSA-N
dichloro-(3-chloropropyl)-methylsilane
82235
≥97.0% (GC)
3-Chloropropyldichloromethylsilane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2047

TSCA : Yes

SDS
Documents

Documents