missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloropropyldichloromethylsilane 97.0+%, TCI America™
Supplier: TCI America D199525ML
Specifications
| 3-Chloropropyldichloromethylsilane | |
| 185°C | |
| MFCD00000996 | |
| 2986 | |
| UCJHMXXKIKBHQP-UHFFFAOYSA-N | |
| dichloro-(3-chloropropyl)-methylsilane | |
| 82235 | |
| ≥97.0% (GC) |
| 7787-93-1 | |
| C4H9Cl3Si | |
| 25 mL | |
| 3-(Dichloromethylsilyl)propyl Chloride | |
| C[Si](CCCCl)(Cl)Cl | |
| 191.551 | |
| 191.55 | |
| Liquid |
Chemical Identifiers
| 7787-93-1 | |
| 191.551 | |
| UCJHMXXKIKBHQP-UHFFFAOYSA-N | |
| 82235 | |
| C[Si](CCCCl)(Cl)Cl |
| C4H9Cl3Si | |
| MFCD00000996 | |
| 3-(Dichloromethylsilyl)propyl Chloride | |
| dichloro-(3-chloropropyl)-methylsilane |
Safety and Handling
EINECSNumber : (2)-2047
TSCA : Yes