Learn More
(3-Chloropropyl)trimethoxysilane, 97+%, packaged under Argon in resealable ChemSeal™ bottles
CAS: 2530-87-2 | C6H15ClO3Si | 198.718 g/mol
Supplier: Thermo Scientific Chemicals 042413AK
Description
(3-Chloropropyl)trimethoxysilane is used as adhesives and sealant chemicals. It is also used in the preparation of 3-iodopropyltrimethoxysilane by reacting with sodium iodide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
| 195°C to 197°C | |
| -50°C | |
| Liquid | |
| C6H15ClO3Si | |
| 969627 | |
| 3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan | |
| OXYZDRAJMHGSMW-UHFFFAOYSA-N | |
| 3-chloropropyl(trimethoxy)silane | |
| 62449 | |
| ≥97% | |
| ChemSeal™ bottles | |
| Moisture sensitive |
| (3-Chloropropyl)trimethoxysilane | |
| 2530-87-2 | |
| 250 mL | |
| MFCD00000997 | |
| 78°C (172°F) | |
| Miscible with organic solvents. | |
| CO[Si](CCCCl)(OC)OC | |
| 198.718 | |
| 198.72 | |
| Recognizable | |
| 1.42 | |
| 1.077 g/mL |
Chemical Identifiers
| 2530-87-2 | |
| 198.718 | |
| OXYZDRAJMHGSMW-UHFFFAOYSA-N | |
| 62449 | |
| CO[Si](CCCCl)(OC)OC |
| C6H15ClO3Si | |
| MFCD00000997 | |
| 3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan | |
| 3-chloropropyl(trimethoxy)silane |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 219-787-9
RTECSNumber : VV2680000
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only