missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)triethoxysilane, 97+%

Catalog No. AC391890250
Change view
Click to view available options
Quantity:
25 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391890250 25 g Glass bottle
AC391890010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391890250 Supplier Thermo Scientific Chemicals Supplier No. 391890250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5089-70-3 | C9H21ClO3Si | 240.8 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5089-70-3
Molecular Formula C9H21ClO3Si
Molecular Weight (g/mol) 240.8
MDL Number MFCD00018985
InChI Key KSCAZPYHLGGNPZ-UHFFFAOYSA-N
Synonym 3-chloropropyl triethoxysilane, 3-chloropropyl triethoxy silane, silane, 3-chloropropyl triethoxy, unii-x7rb20518m, triethoxy gamma-chloropropyl silane, gamma-chloropropyltriethoxysilane, triethoxy .gamma.-chloropropyl silane, chloropropyl triethoxysilane, dynasylan cpteo
PubChem CID 78771
IUPAC Name 3-chloropropyl(triethoxy)silane
SMILES CCO[Si](CCCCl)(OCC)OCC

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Yellow
Density 1.0040g/mL
Boiling Point 230.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Linear Formula Cl(CH2)3Si(OCH2CH3)
Quantity 25 g
Specific Gravity 1.004
Solubility Information Solubility in water: insoluble
Formula Weight 240.8
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material (3-Chloropropyl)triethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-805-6

RUO – Research Use Only