missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyl phenyl sulfide, 97%

CAS: 4911-65-3 | C9H11ClS | 186.70 g/mol

Supplier:  Thermo Scientific Chemicals B2134514

 View more versions of this product

Catalog No. AAB2134514


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloropropyl phenyl sulfide
Stench
MFCD00040122
3-chloropropyl phenyl sulfide, 3-chloropropyl sulfanyl benzene, 3-chloropropyl thio benzene, 3-chloropropylphenylsulfide, acmc-20an5p, 3-phenylthiopropylchloride, 3-phenylthiopropyl chloride, 1-chloro-3-phenylthiopropane, 3-chloro-1-phenylthiopropane
ClCCCSC1=CC=CC=C1
186.70
186.7
4911-65-3
C9H11ClS
25 g
SBRLITLVSMPPFI-UHFFFAOYSA-N
3-chloropropylsulfanylbenzene
269606
97%

Chemical Identifiers

4911-65-3
186.70
SBRLITLVSMPPFI-UHFFFAOYSA-N
269606
ClCCCSC1=CC=CC=C1
C9H11ClS
MFCD00040122
3-chloropropyl phenyl sulfide, 3-chloropropyl sulfanyl benzene, 3-chloropropyl thio benzene, 3-chloropropylphenylsulfide, acmc-20an5p, 3-phenylthiopropylchloride, 3-phenylthiopropyl chloride, 1-chloro-3-phenylthiopropane, 3-chloro-1-phenylthiopropane
3-chloropropylsulfanylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only