missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyl phenyl sulfide, 97%

Catalog No. aab2134506
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAB2134506 5 g
AAB2134514 25 g
2 options

Catalog No. AAB2134506

Supplier: Thermo Scientific Chemicals B2134506

Only null left

CAS: 4911-65-3 | C9H11ClS | 186.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4911-65-3
Molecular Formula C9H11ClS
Molecular Weight (g/mol) 186.70
MDL Number MFCD00040122
InChI Key SBRLITLVSMPPFI-UHFFFAOYSA-N
Synonym 3-chloropropyl phenyl sulfide, 3-chloropropyl sulfanyl benzene, 3-chloropropyl thio benzene, 3-chloropropylphenylsulfide, acmc-20an5p, 3-phenylthiopropylchloride, 3-phenylthiopropyl chloride, 1-chloro-3-phenylthiopropane, 3-chloro-1-phenylthiopropane
PubChem CID 269606
IUPAC Name 3-chloropropylsulfanylbenzene
SMILES ClCCCSC1=CC=CC=C1

Specifications

Odor Stench
Quantity 5 g
Formula Weight 186.7
Percent Purity 97%
Chemical Name or Material 3-Chloropropyl phenyl sulfide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only