missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyl isocyanate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1599306

 View more versions of this product

Catalog No. AAL1599306

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloropropyl isocyanate
1.165
61°C (141°F)
C4H6ClNO
MFCD00010384
UN2206
Moisture sensitive
RQAVSDINDRNIKL-UHFFFAOYSA-N
1-chloro-3-isocyanatopropane
166752
97%
13010-19-0
152°C to 154°C
Pungent
1.452
5 g
1098789
3-chloropropyl isocyanate, 3-chloropropylisocyanate, propane, 1-chloro-3-isocyanato, 1-chloro-3-isocyanato-propane, chloropropylisocyanate, chloropropyl isocyanate, 3-chloropropanisocyanate, chloro propyl isocyanate, acmc-20an5q, 3-chloropropyl-isocyanate
ClCCCN=C=O
119.55
119.55

Chemical Identifiers

13010-19-0
119.55
RQAVSDINDRNIKL-UHFFFAOYSA-N
166752
ClCCCN=C=O
C4H6ClNO
MFCD00010384
3-chloropropyl isocyanate, 3-chloropropylisocyanate, propane, 1-chloro-3-isocyanato, 1-chloro-3-isocyanato-propane, chloropropylisocyanate, chloropropyl isocyanate, 3-chloropropanisocyanate, chloro propyl isocyanate, acmc-20an5q, 3-chloropropyl-isocyanate
1-chloro-3-isocyanatopropane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H334-H318-H227-H315-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.

P210-P235-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H301+H311+H331-H314-H334-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only