missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog No. p-7046713
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1599306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1599306 Supplier Thermo Scientific Chemicals Supplier No. L1599306
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13010-19-0
Molecular Formula C4H6ClNO
Molecular Weight (g/mol) 119.55
MDL Number MFCD00010384
InChI Key RQAVSDINDRNIKL-UHFFFAOYSA-N
Synonym 3-chloropropyl isocyanate, 3-chloropropylisocyanate, propane, 1-chloro-3-isocyanato, 1-chloro-3-isocyanato-propane, chloropropylisocyanate, chloropropyl isocyanate, 3-chloropropanisocyanate, chloro propyl isocyanate, acmc-20an5q, 3-chloropropyl-isocyanate
PubChem CID 166752
IUPAC Name 1-chloro-3-isocyanatopropane
SMILES ClCCCN=C=O

Specifications

Density 1.165
Boiling Point 152°C to 154°C
Flash Point 61°C (141°F)
Odor Pungent
Refractive Index 1.452
Quantity 5 g
UN Number UN2206
Beilstein 1098789
Sensitivity Moisture sensitive
Formula Weight 119.55
Percent Purity 97%
Chemical Name or Material 3-Chloropropyl isocyanate
Show More Show Less
Hazard Category H227-H301+H311+H331-H314-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H318-H227-H315-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only