missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Chloropropyl)diethoxy(methyl)silane 98.0+%, TCI America™
$76.43 - $152.86
Chemical Identifiers
| CAS | 13501-76-3 |
|---|---|
| Molecular Formula | C8H19ClO2Si |
| Molecular Weight (g/mol) | 210.773 |
| MDL Number | MFCD05663910 |
| InChI Key | KEZMLECYELSZDC-UHFFFAOYSA-N |
| PubChem CID | 83536 |
| IUPAC Name | 3-chloropropyl-diethoxy-methylsilane |
| SMILES | CCO[Si](C)(CCCCl)OCC |
Chemical Identifiers
| 13501-76-3 | |
| 210.773 | |
| KEZMLECYELSZDC-UHFFFAOYSA-N | |
| 3-chloropropyl-diethoxy-methylsilane |
| C8H19ClO2Si | |
| MFCD05663910 | |
| 83536 | |
| CCO[Si](C)(CCCCl)OCC |
Specifications
| 13501-76-3 | |
| 109°C | |
| MFCD05663910 | |
| KEZMLECYELSZDC-UHFFFAOYSA-N | |
| 3-chloropropyl-diethoxy-methylsilane | |
| 83536 | |
| ≥98.0% (GC) | |
| (3-Chloropropyl)diethoxy(methyl)silane |
| Colorless | |
| C8H19ClO2Si | |
| 5 mL | |
| CCO[Si](C)(CCCCl)OCC | |
| 210.773 | |
| 210.77 | |
| Liquid |
Safety and Handling
TSCA : No