missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Chloropropyl)diethoxy(methyl)silane 98.0+%, TCI America™
Supplier: TCI America C309225ML
Specifications
| (3-Chloropropyl)diethoxy(methyl)silane | |
| Colorless | |
| C8H19ClO2Si | |
| 25 mL | |
| CCO[Si](C)(CCCCl)OCC | |
| 210.773 | |
| 210.77 | |
| Liquid |
| 13501-76-3 | |
| 109°C | |
| MFCD05663910 | |
| KEZMLECYELSZDC-UHFFFAOYSA-N | |
| 3-chloropropyl-diethoxy-methylsilane | |
| 83536 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 13501-76-3 | |
| 210.773 | |
| KEZMLECYELSZDC-UHFFFAOYSA-N | |
| 3-chloropropyl-diethoxy-methylsilane |
| C8H19ClO2Si | |
| MFCD05663910 | |
| 83536 | |
| CCO[Si](C)(CCCCl)OCC |
Safety and Handling
TSCA : No