missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionitrile 98.0+%, TCI America™

Supplier:  TCI America C0277500ML

 View more versions of this product

Catalog No. C0277500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Chloropropionitrile
Colorless
C3H4ClN
500 mL
3-chloropropionitrile, propanenitrile, 3-chloro, 3-chloropropiononitrile, 1-chloro-2-cyanoethane, 3-chloropropanonitrile, 3-chlorpropannitril, propionitrile, 3-chloro, beta-chloropropionitrile, usaf a-8798, rcra waste number p027
ClCCC#N
89.52
89.52
Liquid
542-76-7
176°C
MFCD00001952
3276
GNHMRTZZNHZDDM-UHFFFAOYSA-N
3-chloropropanenitrile
10963
≥98.0% (GC)

Chemical Identifiers

542-76-7
89.52
GNHMRTZZNHZDDM-UHFFFAOYSA-N
10963
ClCCC#N
C3H4ClN
MFCD00001952
3-chloropropionitrile, propanenitrile, 3-chloro, 3-chloropropiononitrile, 1-chloro-2-cyanoethane, 3-chloropropanonitrile, 3-chlorpropannitril, propionitrile, 3-chloro, beta-chloropropionitrile, usaf a-8798, rcra waste number p027
3-chloropropanenitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2773

RTECSNumber : UG1400000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions