accessibility menu, dialog, popup

UserName

ClN

CAS RN 542-76-7

IUPAC Name: 3-chloropropanenitrile
Synonyms: 3-chloropropionitrile
Molecular Weight (g/mol): 89.52
Molecular Formula: C3H4ClN
InChi Key: GNHMRTZZNHZDDM-UHFFFAOYSA-N
SMILES: ClCCC#N
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)