missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionitrile 98.0+%, TCI America™

Catalog No. C027725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C027725ML 25 mL
C0277500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C027725ML Supplier TCI America Supplier No. C027725ML

Chemical Identifiers

CAS 542-76-7
Molecular Formula C3H4ClN
Molecular Weight (g/mol) 89.52
MDL Number MFCD00001952
InChI Key GNHMRTZZNHZDDM-UHFFFAOYSA-N
Synonym 3-chloropropionitrile, propanenitrile, 3-chloro, 3-chloropropiononitrile, 1-chloro-2-cyanoethane, 3-chloropropanonitrile, 3-chlorpropannitril, propionitrile, 3-chloro, beta-chloropropionitrile, usaf a-8798, rcra waste number p027
PubChem CID 10963
IUPAC Name 3-chloropropanenitrile
SMILES ClCCC#N

Specifications

Color Colorless
Boiling Point 176°C
Quantity 25 mL
UN Number 3276
Formula Weight 89.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropropionitrile
EINECSNumber (2)-2773
RTECSNumber UG1400000
TSCA Yes