missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionitrile, 98%

Catalog No. p-7044327
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0590214 25 g
AAL0590222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0590214 Supplier Thermo Scientific Chemicals Supplier No. L0590214
Only null left

CAS: 542-76-7 | C3H4ClN | 89.52 g/mol

3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.

Solubility
Soluble in water. (4.5g/100mL, 25°C). Miscible in Ether, Alcohols, Benzene, Acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.

Chemical Identifiers

CAS 542-76-7
Molecular Formula C3H4ClN
Molecular Weight (g/mol) 89.52
MDL Number MFCD00001952
InChI Key GNHMRTZZNHZDDM-UHFFFAOYSA-N
Synonym 3-chloropropionitrile, propanenitrile, 3-chloro, 3-chloropropiononitrile, 1-chloro-2-cyanoethane, 3-chloropropanonitrile, 3-chlorpropannitril, propionitrile, 3-chloro, beta-chloropropionitrile, usaf a-8798, rcra waste number p027
PubChem CID 10963
IUPAC Name 3-chloropropanenitrile
SMILES ClCCC#N

Specifications

Melting Point -51°C
Density 1.143
Boiling Point 174°C to 176°C
Flash Point 75°C (167°F)
Refractive Index 1.438
Quantity 25 g
UN Number UN3276
Beilstein 1098454
Merck Index 14,2160
Solubility Information Soluble in water. (4.5g/100mL,25°C). Miscible in Ether,Alcohols,Benzene,Acetone.
Formula Weight 89.53
Percent Purity 98%
Chemical Name or Material 3-Chloropropionitrile
Show More Show Less
Hazard Category H227-H300-H331
Hazard Statement GHS H Statement
H300-H227
Fatal if swallowed.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P304+P340-P311-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S
EINECSNumber 208-827-0
RTECSNumber UG1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only