missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorophenylmagnesium bromide, 1M in MeTHF
Description
3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1'-DiBoc-2,2'-Biaziridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1′-DiBoc-2,2′-Biaziridine.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents. Reacts with water.
3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1′-DiBoc-2,2′-Biaziridine.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents. Reacts with water.
Specifications
Specifications
| Quantity | 50 mL |
| UN Number | UN2924 |
| Formula Weight | 215.76 |
| Concentration or Composition (by Analyte or Components) | 1M in MeTHF |
| Chemical Name or Material | 3-Chlorophenylmagnesium bromide |
Safety and Handling
| Hazard Category | H225-H302-H314-H335 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only