missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylmagnesium bromide, 1M in MeTHF

Catalog No. AAH51161AD
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51161AD 50 mL
AAH51161AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51161AD Supplier Thermo Scientific Chemicals Supplier No. H51161AD

CAS: 36229-42-2 | C6H4BrClMg | 215.76 g/mol

3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1'-DiBoc-2,2'-Biaziridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1′-DiBoc-2,2′-Biaziridine.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents. Reacts with water.

Chemical Identifiers

CAS 36229-42-2
Molecular Formula C6H4BrClMg
Molecular Weight (g/mol) 215.76
MDL Number MFCD00672003
InChI Key ZIYRUWPTUPFRQU-UHFFFAOYSA-M
Synonym 3-chlorophenylmagnesium bromide, 3-chlorophenyl magnesium bromide, magnesium,bromo 3-chlorophenyl, m-chlorophenylmagnesium bromide, m-chlorophenyl magnesium bromide, 3-chloro phenyl magnesium bromide, 3-chloro-phenyl magnesium bromide, ziyruwptupfrqu-uhfffaoysa-m, grignard reagent
PubChem CID 12252967
IUPAC Name magnesium;chlorobenzene;bromide
SMILES ClC1=CC([Mg]Br)=CC=C1

Specifications

Quantity 50 mL
UN Number UN2924
Formula Weight 215.76
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material 3-Chlorophenylmagnesium bromide
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only