missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorophenylhydrazine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America C14775G
Specifications
| 3-Chlorophenylhydrazine Hydrochloride | |
| White-Yellow | |
| MFCD00012935 | |
| 3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride | |
| C1=CC(=CC(=C1)Cl)NN.Cl | |
| 179.044 | |
| 179.04 | |
| Crystalline Powder |
| 2312-23-4 | |
| C6H8Cl2N2 | |
| 5 g | |
| CRRIAWUJYMLJOE-UHFFFAOYSA-N | |
| (3-chlorophenyl)hydrazine;hydrochloride | |
| 75331 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 2312-23-4 | |
| 179.044 | |
| CRRIAWUJYMLJOE-UHFFFAOYSA-N | |
| 75331 | |
| C1=CC(=CC(=C1)Cl)NN.Cl |
| C6H8Cl2N2 | |
| MFCD00012935 | |
| 3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride | |
| (3-chlorophenyl)hydrazine;hydrochloride |
Safety and Handling
TSCA : No