missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylhydrazine hydrochloride, 97%

Catalog No. p-7022140
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1477009 10 g
AAA1477018 50 g
AAA1477030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1477009 Supplier Thermo Scientific Chemicals Supplier No. A1477009

CAS: 2312-23-4 | C6H8Cl2N2 | 179.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2312-23-4
Molecular Formula C6H8Cl2N2
Molecular Weight (g/mol) 179.044
MDL Number MFCD00012935
InChI Key CRRIAWUJYMLJOE-UHFFFAOYSA-N
Synonym 3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride
PubChem CID 75331
IUPAC Name (3-chlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC(=C1)Cl)NN.Cl

Specifications

Melting Point ∼242°C (decomposition)
Odor Odorless
Quantity 10 g
Beilstein 3565828
Formula Weight 179.05
Percent Purity 97%
Chemical Name or Material 3-Chlorophenylhydrazine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 219-005-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only