missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorophenylacetone 95.0+%, TCI America™
$199.46 - $685.97
Chemical Identifiers
| CAS | 14123-60-5 |
|---|---|
| Molecular Formula | C9H9ClO |
| Molecular Weight (g/mol) | 168.62 |
| MDL Number | MFCD00082872 |
| InChI Key | VCNYPJMEQHTAHS-UHFFFAOYSA-N |
| Synonym | 1-3-chlorophenyl propan-2-one, 3-chlorophenylacetone, 1-3-chlorophenyl acetone, 1-3-chlorophenyl-2-propanone, pubchem19805, 3-chlorophenyl-acetone, m-chlorophenyl acetone, 2-propanone,1-3-chlorophenyl, 1-3-chloro-phenyl-propan-2-one, 2-propanone, 1-3-chlorophenyl |
| PubChem CID | 2734097 |
| IUPAC Name | 1-(3-chlorophenyl)propan-2-one |
| SMILES | CC(=O)CC1=CC=CC(Cl)=C1 |
Chemical Identifiers
| 14123-60-5 | |
| 168.62 | |
| VCNYPJMEQHTAHS-UHFFFAOYSA-N | |
| 2734097 | |
| CC(=O)CC1=CC=CC(Cl)=C1 |
| C9H9ClO | |
| MFCD00082872 | |
| 1-3-chlorophenyl propan-2-one, 3-chlorophenylacetone, 1-3-chlorophenyl acetone, 1-3-chlorophenyl-2-propanone, pubchem19805, 3-chlorophenyl-acetone, m-chlorophenyl acetone, 2-propanone,1-3-chlorophenyl, 1-3-chloro-phenyl-propan-2-one, 2-propanone, 1-3-chlorophenyl | |
| 1-(3-chlorophenyl)propan-2-one |
Specifications
| 14123-60-5 | |
| 99°C | |
| MFCD00082872 | |
| 1-3-chlorophenyl propan-2-one, 3-chlorophenylacetone, 1-3-chlorophenyl acetone, 1-3-chlorophenyl-2-propanone, pubchem19805, 3-chlorophenyl-acetone, m-chlorophenyl acetone, 2-propanone,1-3-chlorophenyl, 1-3-chloro-phenyl-propan-2-one, 2-propanone, 1-3-chlorophenyl | |
| CC(=O)CC1=CC=CC(Cl)=C1 | |
| 168.62 | |
| 168.62 | |
| Liquid |
| Yellow | |
| C9H9ClO | |
| 5 g | |
| VCNYPJMEQHTAHS-UHFFFAOYSA-N | |
| 1-(3-chlorophenyl)propan-2-one | |
| 2734097 | |
| ≥95.0% (GC) | |
| 3-Chlorophenylacetone |
Safety and Handling
TSCA : No