missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylacetone 95.0+%, TCI America™

Catalog No. C28955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28955G 5 g
C289525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28955G Supplier TCI America Supplier No. C28955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14123-60-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00082872
InChI Key VCNYPJMEQHTAHS-UHFFFAOYSA-N
Synonym 1-3-chlorophenyl propan-2-one, 3-chlorophenylacetone, 1-3-chlorophenyl acetone, 1-3-chlorophenyl-2-propanone, pubchem19805, 3-chlorophenyl-acetone, m-chlorophenyl acetone, 2-propanone,1-3-chlorophenyl, 1-3-chloro-phenyl-propan-2-one, 2-propanone, 1-3-chlorophenyl
PubChem CID 2734097
IUPAC Name 1-(3-chlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC(Cl)=C1

Specifications

Color Yellow
Boiling Point 99°C
Quantity 5 g
Formula Weight 168.62
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorophenylacetone