missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylacetone, 94%

CAS: 14123-60-5 | C9H9ClO | 168.62 g/mol

$61.46 - $180.27

Chemical Identifiers

CAS 14123-60-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00082872
InChI Key VCNYPJMEQHTAHS-UHFFFAOYSA-N
PubChem CID 2734097
IUPAC Name 1-(3-chlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC(Cl)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350690010
SDS View Documents
Thermo Scientific Chemicals
350690010
1 g Glass bottle
Each for $61.46
Only null left
 
AC350690050
SDS View Documents
Thermo Scientific Chemicals
350690050
5 g Glass bottle
Each for $180.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14123-60-5
168.62
VCNYPJMEQHTAHS-UHFFFAOYSA-N
1-(3-chlorophenyl)propan-2-one
C9H9ClO
MFCD00082872
2734097
CC(=O)CC1=CC=CC(Cl)=C1

Specifications

14123-60-5
Authentic
Glass bottle
1.5320 to 1.5360
1 g
1-3-chlorophenyl propan-2-one, 3-chlorophenylacetone, 1-3-chlorophenyl acetone, 1-3-chlorophenyl-2-propanone, pubchem19805, 3-chlorophenyl-acetone, m-chlorophenyl acetone, 2-propanone,1-3-chlorophenyl, 1-3-chloro-phenyl-propan-2-one, 2-propanone, 1-3-chlorophenyl
CC(=O)CC1=CC=CC(Cl)=C1
168.62
168.62
Liquid
Yellow
92% min. (GC)
C9H9ClO
MFCD00082872
07,IV,691
VCNYPJMEQHTAHS-UHFFFAOYSA-N
1-(3-chlorophenyl)propan-2-one
2734097
94%
3-Chlorophenylacetone
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only